siRNA Details
| Gene Symbol: TTC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114108.1 | NP_001107580.1 | 2, 3 | 736 | ||
| NM_017904.3 | NP_060374.2 | 2, 3 | 736 | |||
| XM_011541671.2 | XP_011539973.1 | 2, 3 | 1847 | |||
| XM_017001582.1 | XP_016857071.1 | 2, 3 | 1870 | |||
| GenBank mRNA | AK000626.1 | - | 2, 3 | 666 | ||
| AL049431.1 | - | 1 | 178 | |||

TTC22
Genomic Map