siRNA Details
| Gene Symbol: Fam110c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025051.1 | NP_001020222.1 | 2 | 1443 | ||
| GenBank mRNA | BC085778.1 | - | 2 | 1443 | 7106593 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam110c| Gene Symbol: Fam110c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025051.1 | NP_001020222.1 | 2 | 1443 | ||
| GenBank mRNA | BC085778.1 | - | 2 | 1443 | 7106593 | |