siRNA Details
| Gene Symbol: Gmppa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025056.1 | NP_001020227.1 | 12 | 1338 | ||
| XM_006245267.2 | XP_006245329.1 | 13 | 1256 | |||
| XM_006245268.2 | XP_006245330.1 | 13 | 1327 | |||
| XM_006245269.2 | XP_006245331.1 | 13 | 1321 | |||
| XM_006245270.2 | XP_006245332.1 | 13 | 1224 | |||
| XM_006245271.2 | XP_006245333.1 | 12 | 1241 | |||
| XM_008767291.1 | XP_008765513.1 | 13 | 1363 | |||
| GenBank mRNA | BC083763.1 | - | 12 | 1338 | 7193128 | |

Gmppa
Genomic Map