siRNA Details
| Gene Symbol: Syce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025058.2 | NP_001020229.2 | 9 | 589 | ||
| XM_006230572.2 | XP_006230634.1 | 9 | 506 | |||
| GenBank mRNA | BC083907.1 | - | 9 | 571 | 7134037 | |
| CK596157.1 | - | 9 | 589 | |||

Syce1
Genomic Map