siRNA Details
| Gene Symbol: Syce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025058.2 | NP_001020229.2 | 13 | 1207 | ||
| XM_006230572.2 | XP_006230634.1 | 13 | 1124 | |||
| GenBank mRNA | BC083907.1 | - | 13 | 1189 | 7134037 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syce1| Gene Symbol: Syce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025058.2 | NP_001020229.2 | 13 | 1207 | ||
| XM_006230572.2 | XP_006230634.1 | 13 | 1124 | |||
| GenBank mRNA | BC083907.1 | - | 13 | 1189 | 7134037 | |