siRNA Details
| Gene Symbol: LOC502684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025060.1 | NP_001020231.1 | 10 | 1098 | ||
| XM_006235246.2 | XP_006235308.1 | 10 | 1161 | |||
| XM_006235247.2 | XP_006235309.1 | 10 | 1159 | |||
| XM_006235248.2 | XP_006235310.1 | 10 | 1260 | |||
| XM_006235249.2 | XP_006235311.1 | 11 | 1138 | |||
| XM_006235251.2 | XP_006235313.1 | 10 | 1138 | |||
| XM_008762376.1 | XP_008760598.1 | 10 | 1239 | |||
| XM_008762377.1 | XP_008760599.1 | 11 | 1198 | |||
| XM_008762378.1 | XP_008760600.1 | 10 | 1140 | |||
| GenBank mRNA | BC083680.1 | - | 10 | 1098 | 7135883 | |

LOC502684
Genomic Map