siRNA Details
| Gene Symbol: Scrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025063.1 | NP_001020234.1 | 8 | 1405 | ||
| XM_006236512.2 | XP_006236574.1 | 8 | 1393 | |||
| XM_008762916.1 | XP_008761138.1 | 8 | 1257 | |||
| GenBank mRNA | BC079152.1 | - | 8 | 1405 | 7128646 | |

Scrn1
Genomic Map