siRNA Details
| Gene Symbol: LOC502894 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025064.1 | NP_001020235.1 | 5 | 1772 | ||
| XM_006237501.2 | XP_006237563.1 | 6 | 1851 | |||
| GenBank mRNA | BC093610.1 | - | 5 | 1772 | 7319640 | |
| FQ217296.1 | - | 6 | 1813 | |||

LOC502894
Genomic Map