siRNA Details
Gene Symbol: Prmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025144.1 | NP_001020315.1 | 5, 6 | 634 | ||
GenBank mRNA | BC079112.1 | - | 5, 6 | 634 | 7125748 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025144.1 | NP_001020315.1 | 5, 6 | 634 | ||
GenBank mRNA | BC079112.1 | - | 5, 6 | 634 | 7125748 |