siRNA Details
Gene Symbol: Tas2r137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025149.1 | NP_001020320.1 | 1 | 793 | ||
XM_006236362.2 | XP_006236424.1 | 2 | 6783 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tas2r137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025149.1 | NP_001020320.1 | 1 | 793 | ||
XM_006236362.2 | XP_006236424.1 | 2 | 6783 |