siRNA Details
| Gene Symbol: CRAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020825.3 | NP_065876.3 | 20 | 6374 | ||
| GenBank mRNA | AB037847.2 | - | 18 | 5767 | ||
| AK056087.1 | - | 1 | 1385 | |||
| BC137172.1 | - | 19 | 5924 | 9021169 | ||
| BC137173.1 | - | 19 | 5924 | 9021170 | ||
| BC152477.1 | - | 18 | 5767 | 8862615 | ||

CRAMP1
Genomic Map