siRNA Details
| Gene Symbol: Fez2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053600.2 | NP_446052.2 | 6 | 970 | ||
| GenBank mRNA | AB076183.1 | - | 6 | 970 | ||
| AF120109.1 | - | 1 | 63 | |||
| CB725415.1 | - | 2 | 212 | |||
| U64689.1 | - | 6 | 796 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fez2| Gene Symbol: Fez2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053600.2 | NP_446052.2 | 6 | 970 | ||
| GenBank mRNA | AB076183.1 | - | 6 | 970 | ||
| AF120109.1 | - | 1 | 63 | |||
| CB725415.1 | - | 2 | 212 | |||
| U64689.1 | - | 6 | 796 | |||