siRNA Details
| Gene Symbol: Defb41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183124.3 | NP_898947.1 | 2 | 236 | ||
| GenBank mRNA | AK020304.1 | - | 2 | 237 | ||
| DQ012027.1 | - | 2 | 236 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Defb41| Gene Symbol: Defb41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183124.3 | NP_898947.1 | 2 | 236 | ||
| GenBank mRNA | AK020304.1 | - | 2 | 237 | ||
| DQ012027.1 | - | 2 | 236 | |||