siRNA Details
| Gene Symbol: Spink8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021276.1 | - | 3, 4 | 367 | ||
| BC048588.1 | - | 3, 4 | 384 | 6707477 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spink8| Gene Symbol: Spink8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021276.1 | - | 3, 4 | 367 | ||
| BC048588.1 | - | 3, 4 | 384 | 6707477 | ||