siRNA Details
| Gene Symbol: Zfp691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145935.1 | NP_001139407.1 | 2 | 282 | ||
| NM_001145936.1 | NP_001139408.1 | 2 | 146 | |||
| NM_183140.3 | NP_898963.2 | 2 | 899 | |||
| GenBank mRNA | AK032431.1 | - | 2 | 901 | ||
| AK170239.1 | - | 2 | 136 | |||
| AK171649.1 | - | 2 | 283 | |||
| BC108395.1 | - | 2 | 133 | 30138157 | ||
| BY233872.1 | - | 2 | 147 | |||

Zfp691
Genomic Map