siRNA Details
| Gene Symbol: Tmem252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183160.3 | NP_898983.1 | 2 | 814 | ||
| GenBank mRNA | AK086895.1 | - | 2 | 816 | ||
| AK142952.1 | - | 2 | 818 | |||
| AK143762.1 | - | 2 | 816 | |||
| BC087877.1 | - | 2 | 817 | 30302311 | ||

Tmem252
Genomic Map