siRNA Details
| Gene Symbol: Rnf182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183204.4 | NP_899027.1 | 2 | 1193 | ||
| GenBank mRNA | AK083175.1 | - | 2 | 1009 | ||
| BC089488.1 | - | 2 | 982 | 30501650 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf182| Gene Symbol: Rnf182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183204.4 | NP_899027.1 | 2 | 1193 | ||
| GenBank mRNA | AK083175.1 | - | 2 | 1009 | ||
| BC089488.1 | - | 2 | 982 | 30501650 | ||