siRNA Details
| Gene Symbol: Pantr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011437.1 | - | 3 | 276 | ||
| AK019154.1 | - | 3 | 276 | |||
| BC058417.1 | - | 3 | 369 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_183252.1 | - | 276 | Chr. - - [-] on Build | ||

Pantr1
Genomic Map