siRNA Details
| Gene Symbol: Smim5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183259.3 | NP_899082.1 | 3 | 713 | ||
| GenBank mRNA | AK004370.1 | - | 3 | 646 | ||
| AK020450.1 | - | 3 | 675 | |||
| BC032884.1 | - | 3 | 891 | |||
| BC065414.1 | - | 4 | 701 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smim5| Gene Symbol: Smim5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183259.3 | NP_899082.1 | 3 | 713 | ||
| GenBank mRNA | AK004370.1 | - | 3 | 646 | ||
| AK020450.1 | - | 3 | 675 | |||
| BC032884.1 | - | 3 | 891 | |||
| BC065414.1 | - | 4 | 701 | |||