siRNA Details
| Gene Symbol: Smco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183283.2 | NP_899106.2 | 3 | 642 | ||
| GenBank mRNA | AK009281.1 | - | 3 | 754 | ||
| BC125555.1 | - | 2 | 600 | 40130070 | ||
| BC131954.1 | - | 2 | 600 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smco1| Gene Symbol: Smco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183283.2 | NP_899106.2 | 3 | 642 | ||
| GenBank mRNA | AK009281.1 | - | 3 | 754 | ||
| BC125555.1 | - | 2 | 600 | 40130070 | ||
| BC131954.1 | - | 2 | 600 | |||