siRNA Details
| Gene Symbol: Ctxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183315.2 | NP_899138.1 | 2 | 1010 | ||
| GenBank mRNA | AK154120.1 | - | 2 | 967 | ||
| AK158759.1 | - | 2 | 904 | |||
| BC028881.1 | - | 2 | 946 | 4224851 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctxn1| Gene Symbol: Ctxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183315.2 | NP_899138.1 | 2 | 1010 | ||
| GenBank mRNA | AK154120.1 | - | 2 | 967 | ||
| AK158759.1 | - | 2 | 904 | |||
| BC028881.1 | - | 2 | 946 | 4224851 | ||