siRNA Details
| Gene Symbol: Khdc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183322.2 | NP_899145.2 | 3 | 1450 | ||
| GenBank mRNA | AK162365.1 | - | 1 | 546 | ||
| AY238603.1 | - | 3 | 1213 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Khdc1a| Gene Symbol: Khdc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183322.2 | NP_899145.2 | 3 | 1450 | ||
| GenBank mRNA | AK162365.1 | - | 1 | 546 | ||
| AY238603.1 | - | 3 | 1213 | |||