siRNA Details
| Gene Symbol: Orai3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198424.3 | NP_940816.1 | 2 | 946 | ||
| GenBank mRNA | AB271216.1 | - | 2 | 772 | ||
| AK037138.1 | - | 3 | 1090 | |||
| AK136955.1 | - | 2 | 947 | |||
| AM712358.1 | - | 2 | 772 | |||
| BC061259.1 | - | 2 | 916 | 30251946 | ||

Orai3
Genomic Map