siRNA Details
| Gene Symbol: Oog2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198661.3 | NP_941063.1 | 5 | 1608 | ||
| GenBank mRNA | AK139660.1 | - | 3 | 2810 | ||
| AY573562.1 | - | 5 | 1608 | |||
| BC052839.1 | - | 5 | 1608 | 30050073 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oog2| Gene Symbol: Oog2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198661.3 | NP_941063.1 | 5 | 1608 | ||
| GenBank mRNA | AK139660.1 | - | 3 | 2810 | ||
| AY573562.1 | - | 5 | 1608 | |||
| BC052839.1 | - | 5 | 1608 | 30050073 | ||