siRNA Details
| Gene Symbol: Cox11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199008.2 | NP_950173.1 | 4 | 1195 | ||
| GenBank mRNA | AK008096.1 | - | 4 | 1209 | ||
| AK143937.1 | - | 4 | 1195 | |||
| AK172067.1 | - | 4 | 1199 | |||
| BC061233.1 | - | 4 | 1206 | 6585499 | ||

Cox11
Genomic Map