siRNA Details
| Gene Symbol: Zfp395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199029.2 | NP_950194.2 | 2 | 289 | ||
| GenBank mRNA | AK155538.1 | - | 2 | 290 | ||
| BC053701.1 | - | 374 | 30054630 | |||
| BY117447.1 | - | 2 | 289 | |||
| BY780785.1 | - | 2 | 239 | |||

Zfp395
Genomic Map