siRNA Details
| Gene Symbol: Rtn4rl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199223.1 | NP_954693.1 | 3 | 982 | ||
| GenBank mRNA | AY250220.1 | - | 3 | 982 | ||
| BC132523.1 | - | 3 | 1046 | 40130800 | ||
| BC138154.1 | - | 3 | 1046 | 8861173 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rtn4rl2| Gene Symbol: Rtn4rl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199223.1 | NP_954693.1 | 3 | 982 | ||
| GenBank mRNA | AY250220.1 | - | 3 | 982 | ||
| BC132523.1 | - | 3 | 1046 | 40130800 | ||
| BC138154.1 | - | 3 | 1046 | 8861173 | ||