siRNA Details
| Gene Symbol: Kcnk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199251.1 | NP_954859.1 | 2 | 988 | ||
| GenBank mRNA | AB052939.1 | - | 2 | 988 | ||
| BC138133.1 | - | 2 | 780 | 8861152 | ||
| BC138134.1 | - | 2 | 780 | 8861153 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnk12| Gene Symbol: Kcnk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199251.1 | NP_954859.1 | 2 | 988 | ||
| GenBank mRNA | AB052939.1 | - | 2 | 988 | ||
| BC138133.1 | - | 2 | 780 | 8861152 | ||
| BC138134.1 | - | 2 | 780 | 8861153 | ||