siRNA Details
| Gene Symbol: BANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083907.2 | NP_001077376.2 | 2 | 212 | ||
| NM_017935.4 | NP_060405.4 | 2 | 443 | |||
| XM_017008337.1 | XP_016863826.1 | 2 | 190 | |||
| GenBank mRNA | AB063170.1 | - | 1 | 79 | ||
| AK000713.1 | - | 2 | 159 | |||
| AK091523.1 | - | 6 | 544 | |||
| AK292133.1 | - | 2 | 287 | |||
| AL834291.1 | - | 2 | 262 | |||
| BC032241.1 | - | 2 | 311 | 5214147 | ||
| DA109829.1 | - | 2 | 441 | |||
| DA946668.1 | - | 2 | 213 | |||

BANK1
Genomic Map