siRNA Details
| Gene Symbol: C2cd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199391.1 | NP_955423.1 | 9 | 1264 | ||
| XM_008768572.1 | XP_008766794.1 | 9 | 1371 | |||
| XM_008768573.1 | XP_008766795.1 | 9 | 1368 | |||
| XM_008768574.1 | XP_008766796.1 | 9 | 1374 | |||
| XM_008768575.1 | XP_008766797.1 | 9 | 1374 | |||
| XM_008768576.1 | XP_008766798.1 | 10 | 973 | |||
| XM_008768577.1 | XP_008766799.1 | 9 | 848 | |||
| XM_008768578.1 | XP_008766800.1 | 9 | 1211 | |||
| GenBank mRNA | BC062389.1 | - | 9 | 1264 | 5620907 | |

C2cd2
Genomic Map