siRNA Details
| Gene Symbol: HEATR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182922.2 | NP_891552.1 | 3 | 501 | ||
| XM_005256013.2 | XP_005256070.1 | 2 | 316 | |||
| XM_011523188.2 | XP_011521490.1 | 3 | 391 | |||
| XM_017023385.1 | XP_016878874.1 | 2 | 564 | |||
| XM_017023386.1 | XP_016878875.1 | 2 | 369 | |||
| GenBank mRNA | AF462442.1 | - | 2 | 308 | ||
| AK000725.1 | - | 2 | 308 | |||
| AK289949.1 | - | 3 | 466 | |||
| BC014415.1 | - | 478 | ||||
| BC018730.1 | - | 3 | 466 | 4907047 | ||
| BC033077.1 | - | 3 | 367 | 4301559 | ||

HEATR3
Genomic Map