siRNA Details
| Gene Symbol: Olfr594 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207143.1 | NP_997026.1 | 1 | 646 | ||
| GenBank mRNA | BC141016.1 | - | 1 | 723 | 9055628 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr594| Gene Symbol: Olfr594 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207143.1 | NP_997026.1 | 1 | 646 | ||
| GenBank mRNA | BC141016.1 | - | 1 | 723 | 9055628 | |