siRNA Details
| Gene Symbol: Olfr813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207147.1 | NP_997030.1 | 1 | 903 | ||
| GenBank mRNA | BC127959.1 | - | 1 | 907 | 40134789 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr813| Gene Symbol: Olfr813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207147.1 | NP_997030.1 | 1 | 903 | ||
| GenBank mRNA | BC127959.1 | - | 1 | 907 | 40134789 | |