siRNA Details
| Gene Symbol: SLC35A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017945.3 | NP_060415.1 | 4 | 558 | ||
| XM_011512948.1 | XP_011511250.1 | 4 | 408 | |||
| GenBank mRNA | AK000737.1 | - | 4 | 461 | ||
| AK098839.1 | - | 4 | 471 | |||
| AK297411.1 | - | 4 | 458 | |||
| AY358994.1 | - | 4 | 467 | |||
| BC005207.1 | - | 4 | 520 | 3829330 | ||
| BC010307.1 | - | 4 | 520 | 3450563 | ||
| BC013046.1 | - | 4 | 462 | 3875085 | ||

SLC35A5
Genomic Map