siRNA Details
Gene Symbol: Ube2dnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276396.1 | NP_001263325.1 | 2 | 499 | ||
GenBank mRNA | BC048523.1 | - | 2 | 499 |
Gene Symbol: Ube2dnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081661.1 | NP_001075130.1 | 2 | 501 | ||
GenBank mRNA | AK015883.1 | - | 2 | 501 |
