siRNA Details
| Gene Symbol: Ube2dnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276396.1 | NP_001263325.1 | 2 | 730 | ||
| GenBank mRNA | BC048523.1 | - | 2 | 730 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2dnl1| Gene Symbol: Ube2dnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276396.1 | NP_001263325.1 | 2 | 730 | ||
| GenBank mRNA | BC048523.1 | - | 2 | 730 | ||