siRNA Details
| Gene Symbol: Olr855 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000409.1 | NP_001000409.1 | 1 | 774 | ||
| XM_006238655.2 | XP_006238717.1 | 2 | 986 | |||
| Gene Symbol: LOC100910606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750015.3 | XP_003750063.1 | 2 | 986 | ||

Olr855
Genomic Map