siRNA Details
| Gene Symbol: Olr858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000584.1 | NP_001000584.1 | 211 | |||
| Gene Symbol: LOC100912491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750009.1 | XP_003750057.1 | 1 | 211 | ||

Olr858
Genomic Map