siRNA Details
| Gene Symbol: NRDE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017970.3 | NP_060440.2 | 7, 8 | 1781 | ||
| XM_011536886.1 | XP_011535188.1 | 8, 9 | 1609 | |||
| XM_017021394.1 | XP_016876883.1 | 8, 9 | 1609 | |||
| XM_017021395.1 | XP_016876884.1 | 8, 9 | 1730 | |||
| XM_017021396.1 | XP_016876885.1 | 7, 8 | 1324 | |||
| GenBank mRNA | AK000870.1 | - | 7, 8 | 1579 | ||
| AK024113.1 | - | 2, 3 | 322 | |||
| AK294958.1 | - | 7, 8 | 1585 | |||
| BC098568.1 | - | 7, 8 | 1559 | 30706812 | ||
| BC118673.1 | - | 4, 5 | 1114 | 40028659 | ||

NRDE2
Genomic Map