siRNA Details
Gene Symbol: Olr1687 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000508.1 | NP_001000508.1 | 1 | 792 | ||
GenBank mRNA | AF091563.1 | - | 1 | 591 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr1687 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000508.1 | NP_001000508.1 | 1 | 792 | ||
GenBank mRNA | AF091563.1 | - | 1 | 591 |