siRNA Details
| Gene Symbol: Olr1250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000806.1 | NP_001000806.1 | 1 | 814 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1250| Gene Symbol: Olr1250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000806.1 | NP_001000806.1 | 1 | 814 | ||