siRNA Details
| Gene Symbol: TMEM248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017994.4 | NP_060464.1 | 2, 3 | 431 | ||
| XM_005250482.3 | XP_005250539.1 | 2, 3 | 283 | |||
| GenBank mRNA | AK000961.1 | - | 2, 3 | 417 | ||
| AK222774.1 | - | 2, 3 | 431 | |||
| BC008675.1 | - | 2, 3 | 411 | 3853834 | ||
| BC010519.1 | - | 2, 3 | 385 | 4150551 | ||
| BC012562.1 | - | 2, 3 | 426 | 4332094 | ||
| BC041764.2 | - | 2, 3 | 400 | 4152659 | ||

TMEM248
Genomic Map