siRNA Details
| Gene Symbol: Olr17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000937.1 | NP_001000937.1 | 305 | |||
| Gene Symbol: LOC100909935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748915.2 | XP_003748963.1 | 1 | 319 | ||

Olr17
Genomic Map