siRNA Details
Gene Symbol: Olr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000937.1 | NP_001000937.1 | 737 |
Gene Symbol: LOC100909935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748915.2 | XP_003748963.1 | 1 | 751 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
