siRNA Details
| Gene Symbol: Olr339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000954.1 | NP_001000954.1 | 1 | 483 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr339| Gene Symbol: Olr339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000954.1 | NP_001000954.1 | 1 | 483 | ||