siRNA Details
Gene Symbol: Olr1256 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001086.1 | NP_001001086.1 | 1 | 121 |
Gene Symbol: LOC100909501 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762441.1 | XP_008760663.1 | 1 | 121 |
Gene Symbol: LOC100912371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749667.3 | XP_003749715.1 | 1 | 121 |
