siRNA Details
| Gene Symbol: C1orf123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304759.1 | NP_001291688.1 | 7 | 450 | ||
| NM_001304760.1 | NP_001291689.1 | 6 | 366 | |||
| NM_017887.2 | NP_060357.1 | 8 | 507 | |||
| GenBank mRNA | AK000587.1 | - | 8 | 485 | ||
| AK092734.1 | - | 6 | 1351 | |||
| AK094371.1 | - | 3 | 1683 | |||
| BC010908.1 | - | 8 | 489 | 4093763 | ||
| BE889129.1 | - | 5 | 319 | |||
| BI603716.1 | - | 7 | 443 | |||
| CR457246.1 | - | 7 | 443 | |||

C1orf123
Genomic Map