siRNA Details
Gene Symbol: Vmac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001720.1 | NP_001001720.1 | 1 | 47 | ||
XM_008766765.1 | XP_008764987.1 | 2 | 147 | |||
XM_008766766.1 | XP_008764988.1 | 1 | 160 | |||
GenBank mRNA | AY521229.1 | - | 1 | 47 |
