siRNA Details
| Gene Symbol: Tex24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013609.2 | NP_001013627.1 | 3 | 1722 | ||
| GenBank mRNA | AK007143.1 | - | 1 | 580 | ||
| AY552600.1 | - | 3 | 1721 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex24| Gene Symbol: Tex24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013609.2 | NP_001013627.1 | 3 | 1722 | ||
| GenBank mRNA | AK007143.1 | - | 1 | 580 | ||
| AY552600.1 | - | 3 | 1721 | |||