siRNA Details
Gene Symbol: LURAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013615.2 | NP_001013633.1 | 2 | 1552 | ||
GenBank mRNA | AK057892.1 | - | 2 | 1552 | ||
CR746264.1 | - | 1 | 113 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LURAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013615.2 | NP_001013633.1 | 2 | 1552 | ||
GenBank mRNA | AK057892.1 | - | 2 | 1552 | ||
CR746264.1 | - | 1 | 113 |